Learn More
Medchemexpress LLC Clofazimine-d7 | 2030-63-9 | 99.0% | 1 MG
SDP
Catalog No. 5000412414
Encompass_Preferred
Clofazimine-d7 is a deuterium-labeled form of Clofazimine, an iminophenazine dye known for its anti-inflammatory effects. It has been used with other antimycobacterial agents to treat conditions like AIDS and Crohn's disease. This compound can be utilized as a tracer or as an internal standard for quantitative analysis through methods such as NMR, GC-MS, or LC-MS.
  • Deuterium-labeled compound
  • Tracer for quantitative analysis
  • Internal standard for analytical methods
  • Used in drug development for pharmacokinetic studies
Catalog No. 50-004-12414 Supplier Medchemexpress LLC Supplier No. HYB1046S1MG
Add to Cart
Add to Cart
Clofazimine-d7 is a deuterium-labeled form of Clofazimine, an iminophenazine dye known for its anti-inflammatory effects. It has been used with other antimycobacterial agents to treat conditions like AIDS and Crohn's disease. This compound can be utilized as a tracer or as an internal standard for quantitative analysis through methods such as NMR, GC-MS, or LC-MS.
  • Deuterium-labeled compound
  • Tracer for quantitative analysis
  • Internal standard for analytical methods
  • Used in drug development for pharmacokinetic studies