Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Clofenapate methyl ester; MCP | 21340-68-1 | 99.0% | 304.77 | 1 ML
SDP

Catalog No. 5000391208
Encompass_Preferred

Methyl clofenapate (Clofenapate methyl ester; MCP) is a peroxisome proliferator that can induce hypolipidemia, peroxisome proliferation, and hepatocarcinogenesis. This compound appears as a white to off-white solid. It has been tested and complies with given specifications.

  • Appearance: White to off-white solid
  • Storage conditions for powder: -20°C for 3 years, 4°C for 2 years
  • Storage conditions in solvent: -80°C for 6 months, -20°C for 1 month
  • 1H NMR spectrum consistent with structure
  • LCMS consistent with structure
  • For research use only

Catalog No. 50-003-91208 Supplier Medchemexpress LLC Supplier No. HY16408210MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl clofenapate (Clofenapate methyl ester; MCP) is a peroxisome proliferator that can induce hypolipidemia, peroxisome proliferation, and hepatocarcinogenesis. This compound appears as a white to off-white solid. It has been tested and complies with given specifications.

  • Appearance: White to off-white solid
  • Storage conditions for powder: -20°C for 3 years, 4°C for 2 years
  • Storage conditions in solvent: -80°C for 6 months, -20°C for 1 month
  • 1H NMR spectrum consistent with structure
  • LCMS consistent with structure
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.