Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Clofencet 250mg | 129025-54-3 | 278.69 g·mol⁻¹ | C13H11ClN2O3 | 250 MG
SDP

Catalog No. 5000226854
Encompass_Preferred

Clofencet is a wheat hybridizing agent used in agricultural research to produce hybrid wheat seeds by inhibiting pollen development without affecting female fertility. It is supplied for research use only.

  • Used to induce wheat hybridization by inhibiting pollen development.
  • High purity: 98.62%.
  • Molecular formula C13H11ClN2O3; molecular weight 278.69 g·mol⁻¹.
  • Light yellow to brown solid; soluble in DMSO (5.83 mg/mL), insoluble in water (<0.1 mg/mL).
  • Available in multiple package sizes including 250 mg.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • For research use only.

Catalog No. 50-002-26854 Supplier Medchemexpress LLC Supplier No. HY114857250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Clofencet is a wheat hybridizing agent used in agricultural research to produce hybrid wheat seeds by inhibiting pollen development without affecting female fertility. It is supplied for research use only.

  • Used to induce wheat hybridization by inhibiting pollen development.
  • High purity: 98.62%.
  • Molecular formula C13H11ClN2O3; molecular weight 278.69 g·mol⁻¹.
  • Light yellow to brown solid; soluble in DMSO (5.83 mg/mL), insoluble in water (<0.1 mg/mL).
  • Available in multiple package sizes including 250 mg.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.