Learn More
Cayman Chemical ClofentezIn 100mg
SDP
Catalog No. 502933490
Encompass_Preferred
An acaricide; has broad-spectrum activity against various plant-feeding mite species; lethal to wild-type, but not chs1 mutant, T. urticae mite eggs (LC50s = 1.2 and >5,000 mg/L, respectively); induces formation of thyroid tumors in male rats from 13-1,000 mg/kg per day
Catalog No. 50-293-3490 Supplier Cayman Chemical Supplier No. 24228100 MG
An acaricide; has broad-spectrum activity against various plant-feeding mite species; lethal to wild-type, but not chs1 mutant, T. urticae mite eggs (LC50s = 1.2 and >5,000 mg/L, respectively); induces formation of thyroid tumors in male rats from 13-1,000 mg/kg per day