Learn More
Enzo Life Sciences Clofibrate (1g). CAS: 637-07-0
SDP
Supplier:  Enzo Life Sciences ALX270256G001
Encompass_Preferred
Activates PPARα. Induces cytochrome P450 4A1 and 4A3 which catalize the omega-oxidation of fatty acids and prostaglandins. Alternative name: 2-(p-Chlorophenoxy)-2-methylpropionic acid ethyl ester. Formula: C12H15ClO3. MW: 242.7. MI: 14: 2377. Purity: ≥98% (GC). Appearance: Colorless oil. Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (25mg/ml). Long Term Storage: Ambient.
Catalog No. 50-200-8449