Learn More
Apexbio Technology LLC Clofibrate 637-07-0 10g
SDP
Catalog No. 5000089005
Encompass_Preferred
Clofibrate (CAS 637-07-0) is a small-molecule agonist targeting peroxisome proliferator-activated receptor alpha (PPAR-alpha) It is designed to activate PPAR-alpha thereby regulating lipid and lipoprotein metabolism pathways Clofibrate exerts its biological activity primarily through PPAR-alpha activation leading to enhanced peroxisome proliferation and fatty acid oxidation In both human and mouse models Clofibrate demonstrates agonist activity with EC50 values of 55 M and 500 M respectively Based on these pharmacological properties Clofibrate holds research potential in investigating lipid metabolism triglyceride regulation and the roles of PPAR-alpha signaling in hyperlipidemia and related metabolic disorders
Catalog No. 50-000-89005 Supplier Apexbio Technology LLC Supplier No. B361010000
Clofibrate (CAS 637-07-0) is a small-molecule agonist targeting peroxisome proliferator-activated receptor alpha (PPAR-alpha) It is designed to activate PPAR-alpha thereby regulating lipid and lipoprotein metabolism pathways Clofibrate exerts its biological activity primarily through PPAR-alpha activation leading to enhanced peroxisome proliferation and fatty acid oxidation In both human and mouse models Clofibrate demonstrates agonist activity with EC50 values of 55 M and 500 M respectively Based on these pharmacological properties Clofibrate holds research potential in investigating lipid metabolism triglyceride regulation and the roles of PPAR-alpha signaling in hyperlipidemia and related metabolic disorders