Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clofibric acid, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111015744 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5744 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5744 Supplier MilliporeSigma Supelco Supplier No. 90323100MG
Add to Cart
Add to Cart
Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 882-09-7
Physical Form Neat
Linear Formula ClC6H4 OC(CH3)2 CO2 H
MDL Number MFCD00004192
Melting Point 120°C to 122°C
Synonym 2-(p-Chlorophenoxy)-2-methylpropionic acid; alpha-(p-Chlorophenoxy)isobutyric acid; Clofibric acid
Type Analytical Standard
Quantity 100 mg
Formula Weight 214.65
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.