Learn More
Cayman Chemical ClofIbrIc AcId-d4 10mg
SDP
Supplier:  Cayman Chemical 3361210 MG
Encompass_Preferred
An internal standard for the quantification of clofibric acid by GC- or LC-MS
Catalog No. 50-294-9077