Learn More
Cayman Chemical ClofIbrIc AcId-d4 1mg
SDP
Supplier:  Cayman Chemical 336121 MG
Encompass_Preferred
An internal standard for the quantification of clofibric acid by GC- or LC-MS
Catalog No. 50-294-8826