Learn More
Cayman Chemical ClofIbrIc AcId-d4 500ug
SDP
Catalog No. 502948572
Encompass_Preferred
An internal standard for the quantification of clofibric acid by GC- or LC-MS
Catalog No. 50-294-8572 Supplier Cayman Chemical Supplier No. 33612500 UG
An internal standard for the quantification of clofibric acid by GC- or LC-MS