Learn More
Eurofins Discoverx Clofilium, 10 mg. (92-1302)
Supplier:  Eurofins Discoverx 921302
Encompass
Clofilium tosylate or 4-Chloro-N,N-diethyl-N-heptylbenzenebutanaminium tosylate, is a K+ channel blocker and functions as a cardiac depressant and anti-arrhythmic agent. Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9818