Learn More
FUJIFILM Biosciences Clomeprop Standard
Catalog No. 5000450406
Encompass_Preferred
Standard material of Clomeprop for pesticide residue analysis Clomeprop standard with 98% purity essential for accurate quantification in pesticide residue analysis Provides reliable performance for regulatory and research settings
Catalog No. 50-004-50406 Supplier FUJIFILM Biosciences Supplier No. 03320731
Standard material of Clomeprop for pesticide residue analysis Clomeprop standard with 98% purity essential for accurate quantification in pesticide residue analysis Provides reliable performance for regulatory and research settings