Learn More
Medchemexpress LLC HY-129105 100mg Medchemexpress, Clomethiazole CAS:533-45-9 Purity:>98%
SDP
Catalog No. 501969796
Encompass_Preferred
Medchemexpress, HY-129105 100mg Clomethiazole CAS:533-45-9 Chlormethiazole is an potent and orally active GABAA agonist. Chlormethiazole inhibits cytochrome P450 isoforms: CYP2A6 and CYP2E1 in human liver microsomes. Chlormethiazole is an anticonvulsant agent and has the potential for treating convulsive status epilepticus. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9796 Supplier Medchemexpress LLC Supplier No. HY129105100MG
Medchemexpress, HY-129105 100mg Clomethiazole CAS:533-45-9 Chlormethiazole is an potent and orally active GABAA agonist. Chlormethiazole inhibits cytochrome P450 isoforms: CYP2A6 and CYP2E1 in human liver microsomes. Chlormethiazole is an anticonvulsant agent and has the potential for treating convulsive status epilepticus. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.