Learn More
Selleck Chemical LLC Clomifene citrate S2561-1g
SDP
Catalog No. 503111641
Encompass_Preferred
Clomifene citrate (NSC 35770 Clomiphene citrate) is a selective estrogen receptor modulator used in the treatment of ovulation induction
Catalog No. 50-311-1641 Supplier Selleck Chemical LLC Supplier No. S25611G
Clomifene citrate (NSC 35770 Clomiphene citrate) is a selective estrogen receptor modulator used in the treatment of ovulation induction