Learn More
Apexbio Technology LLC Clomiphene citrate 50-41-9 5g
SDP
Supplier:  Apexbio Technology LLC B15045000
Encompass_Preferred
Clomiphene citrate (CAS 50-41-9) is a small-molecule modulator targeting estrogen receptors (ER) It is designed to selectively modulate ER-mediated signaling thereby influencing estrogen-dependent cellular pathways Clomiphene citrate exerts its biological activity primarily through competitive inhibition at estrogen receptors It demonstrates both antiestrogenic and estrogenic activities depending on tissue context Based on these pharmacological properties clomiphene citrate holds research potential in the investigation of estrogen receptor-mediated transcriptional regulation cellular proliferation and differentiation as well as hypothalamic-pituitary-gonadal axis regulation and ovarian follicular maturation in reproductive biology studies
Catalog No. 50-000-89997