Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clomiphene citrate salt, MP Biomedicals™

Catalog No. ICN19017410 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19017410 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19017410 Supplier MP Biomedicals, Inc Supplier No. 0219017410
Add to Cart
Add to Cart

Chemical Identifiers

CAS 50-41-9
Molecular Formula C32H36ClNO8
Molecular Weight (g/mol) 598.089
InChI Key PYTMYKVIJXPNBD-BTKVJIOYSA-N
Synonym clomifene citrate, clomiphene citrate, clomid, cis-clomiphene citrate, clomiphene a citrate, zuclomid, chloramiphene, milophene, serophene, z-clomiphene citrate
PubChem CID 6420009
IUPAC Name 2-[4-[(E)-2-chloro-1,2-diphenylethenyl]phenoxy]-N,N-diethylethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES CCN(CC)CCOC1=CC=C(C=C1)C(=C(C2=CC=CC=C2)Cl)C3=CC=CC=C3.C(C(=O)O)C(CC(=O)O)(C(=O)O)O

Specifications

Color White to Yellow
Quantity 10 g
Formula Weight 598.1
Physical Form Crystalline Powder
Chemical Name or Material Clomiphene citrate salt
Recommended Storage Store at 2 to 8°C
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.