Learn More
AdipoGen Clomiphene citrate (1 g)
SDP
Supplier:  AdipoGen CDXC0696G001
Encompass_Preferred
Chemical. CAS: 50-41-9. Formula: C26H28ClNO . C6H8O7. MW: 598.08. Clomiphene is a nonsteroidal triphenylethylene derivative. It acts as a selective estrogen receptor modulator (SERM), that impairs the activation of estrogen receptors (ERs) by 17beta-estradiol, similar to tamoxifen and raloxifene. It potently binds both ERalpha and ERbeta (Ki = 0.9 and 1.2nM, respectively). Clomiphene enhances the release of gonadotropin-releasing hormone, stimulating the release of follicle-stimulating hormone and luteinizing hormone, culminating in ovulation. Triphenylethylene antiestrogens may also be used as antileukemic drugs which kill cells by apoptosis mediated by oxidative stress and activation of PKC.
Catalog No. 50-205-4349