Learn More
Medchemexpress LLC Clomipramine-d3 hydrochloride | 1398065-86-5 | 100.0% | 1 MG
SDP
Catalog No. 5000408849
Encompass_Preferred
Clomipramine-d3 (hydrochloride) is a deuterium-labeled variant of Clomipramine hydrochloride. Clomipramine hydrochloride functions as a blocker for the serotonin transporter (SERT), norepinephrine transporter (NET), and dopamine transporter (DAT), with Ki values of 0.14, 54, and 3 nM, respectively.
  • Used as a tracer.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-08849 Supplier Medchemexpress LLC Supplier No. HYB0457S1MG
Add to Cart
Add to Cart
Clomipramine-d3 (hydrochloride) is a deuterium-labeled variant of Clomipramine hydrochloride. Clomipramine hydrochloride functions as a blocker for the serotonin transporter (SERT), norepinephrine transporter (NET), and dopamine transporter (DAT), with Ki values of 0.14, 54, and 3 nM, respectively.
  • Used as a tracer.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.