Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Clomipramine-d3 hydrochloride | 1398065-86-5 | 100.0% | 10 MG
SDP

Catalog No. 5000298004
Encompass_Preferred

Clomipramine-d3 hydrochloride is the deuterium-labeled hydrochloride salt of clomipramine supplied as a high-purity solid for use as an analytical standard and stable-isotope tracer in research applications such as mass spectrometry, pharmacokinetics, and isotope-dilution assays.

  • Deuterated internal standard for mass spectrometry and isotope-dilution workflows.
  • Hydrochloride salt form supplied as a solid, white to off-white.
  • High purity suitable for analytical applications (99.98% reported).
  • Molecular weight 354.33; formula C19H21D3Cl2N2.
  • Packaged as 10 MG; store at -20°C, sealed, protect from moisture.

Catalog No. 50-002-98004 Supplier Medchemexpress LLC Supplier No. HYB0457S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Clomipramine-d3 hydrochloride is the deuterium-labeled hydrochloride salt of clomipramine supplied as a high-purity solid for use as an analytical standard and stable-isotope tracer in research applications such as mass spectrometry, pharmacokinetics, and isotope-dilution assays.

  • Deuterated internal standard for mass spectrometry and isotope-dilution workflows.
  • Hydrochloride salt form supplied as a solid, white to off-white.
  • High purity suitable for analytical applications (99.98% reported).
  • Molecular weight 354.33; formula C19H21D3Cl2N2.
  • Packaged as 10 MG; store at -20°C, sealed, protect from moisture.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.