Learn More
Medchemexpress LLC Clomipramine-d6 hydrochloride | 1189882-28-7 | 1 MG
SDP
Catalog No. 5000407724
Encompass_Preferred
Clomipramine-d6 hydrochloride is a deuterium labeled form of Clomipramine hydrochloride, a potent 5-HT reuptake blocker with an IC50 value of 1.5 nM. It is a tricyclic antidepressant used in research for depression and obsessive compulsive disorder (OCD). Deuteration can influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs.
  • Suitable for research of depression and obsessive compulsive disorder (OCD).
Catalog No. 50-004-07724 Supplier Medchemexpress LLC Supplier No. HYB0457S11MG
Clomipramine-d6 hydrochloride is a deuterium labeled form of Clomipramine hydrochloride, a potent 5-HT reuptake blocker with an IC50 value of 1.5 nM. It is a tricyclic antidepressant used in research for depression and obsessive compulsive disorder (OCD). Deuteration can influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs.
  • Suitable for research of depression and obsessive compulsive disorder (OCD).