Learn More
Selleck Chemical LLC Clomipramine HCl 50mg 17321-77-6
SDP
Catalog No. 501362533
Encompass_Preferred
Clomipramine HCl is a hydrochloride salt of clomipramine which is a serotonin transporter (SERT), norepinephrine transporter (NET) dopamine transporter (DAT) blocker with Ki of 0.14, 54 and 3 nM, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2533 Supplier Selleck Chemical LLC Supplier No. S254150MG
Clomipramine HCl is a hydrochloride salt of clomipramine which is a serotonin transporter (SERT), norepinephrine transporter (NET) dopamine transporter (DAT) blocker with Ki of 0.14, 54 and 3 nM, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.