Learn More
Medchemexpress LLC Clomipramine hydrochloride | 17321-77-6 | 99.31% | 100 MG
SDP
Catalog No. 5000405306
Encompass_Preferred
Clomipramine hydrochloride is an analytical standard of Clomipramine hydrochloride. It is intended for research and analytical applications. It is a potent 5-HT reuptake blocker with an IC50 value of 1.5 nM and a tricyclic antidepressant used for the research of depression and obsessive-compulsive disorder (OCD). This analytical standard is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS. Its molecular weight is 351.31 and its formula is C19H24Cl2N2. It appears as a solid, white to off-white in color.
  • Analytical standard for research and analytical applications
  • Potent 5-HT reuptake blocker with an IC50 value of 1.5 nM
  • Tricyclic antidepressant for research of depression and obsessive-compulsive disorder (OCD)
  • Suitable for HPLC, GC, and MS experiments
  • White to off-white solid appearance
Catalog No. 50-004-05306 Supplier Medchemexpress LLC Supplier No. HYB0457R100MG
Clomipramine hydrochloride is an analytical standard of Clomipramine hydrochloride. It is intended for research and analytical applications. It is a potent 5-HT reuptake blocker with an IC50 value of 1.5 nM and a tricyclic antidepressant used for the research of depression and obsessive-compulsive disorder (OCD). This analytical standard is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS. Its molecular weight is 351.31 and its formula is C19H24Cl2N2. It appears as a solid, white to off-white in color.
  • Analytical standard for research and analytical applications
  • Potent 5-HT reuptake blocker with an IC50 value of 1.5 nM
  • Tricyclic antidepressant for research of depression and obsessive-compulsive disorder (OCD)
  • Suitable for HPLC, GC, and MS experiments
  • White to off-white solid appearance