Learn More
Medchemexpress LLC Clomipramine hydrochloride | 17321-77-6 | >99.5% | 50 MG
Supplier: Medchemexpress LLC HYB0457R50MG
Clomipramine (Chlorimipramine) hydrochloride is a potent 5-HT reuptake blocker with an IC50 value of 1.5 nM. It is a tricyclic antidepressant suitable for the research of depression and obsessive compulsive disorder (OCD).
- Clomipramine hydrochloride can inhibit reuptake of both noradrenaline and 5-HT, with a stronger inhibition on 5-HT reuptake.
- It interferes with autophagic flux and severely compromises the viability of tumorigenic cells upon cytotoxic stress.
- Clomipramine hydrochloride reduces immobility in the forced swimming test, which is a behavioral model for antidepressants.
- It also inhibits the OCD animal model, marble burying behavior in mice.