Learn More
Medchemexpress LLC Clomipramine hydrochloride | 17321-77-6 | 99.6% | 100 MG
SDP
Catalog No. 5000405305
Encompass_Preferred
Clomipramine hydrochloride is a potent 5-HT reuptake blocker with an IC50 value of 1.5 nM. It is a tricyclic antidepressant primarily used for research into depression and obsessive compulsive disorder (OCD). This compound also inhibits the reuptake of both noradrenaline and 5-HT, with a stronger inhibition of 5-HT reuptake.
  • Potent 5-HT reuptake blocker
  • Tricyclic antidepressant
  • Supports research on depression and obsessive compulsive disorder (OCD)
  • Inhibits both noradrenaline and 5-HT reuptake
  • Interferes with autophagic flux
Catalog No. 50-004-05305 Supplier Medchemexpress LLC Supplier No. HYB0457100MG
Clomipramine hydrochloride is a potent 5-HT reuptake blocker with an IC50 value of 1.5 nM. It is a tricyclic antidepressant primarily used for research into depression and obsessive compulsive disorder (OCD). This compound also inhibits the reuptake of both noradrenaline and 5-HT, with a stronger inhibition of 5-HT reuptake.
  • Potent 5-HT reuptake blocker
  • Tricyclic antidepressant
  • Supports research on depression and obsessive compulsive disorder (OCD)
  • Inhibits both noradrenaline and 5-HT reuptake
  • Interferes with autophagic flux