Learn More
Medchemexpress LLC Clomipramine hydrochloride | 17321-77-6 | 99.6% | 500 MG
SDP
Catalog No. 5000408419
Encompass_Preferred
Clomipramine hydrochloride is a potent 5-HT reuptake blocker with an IC50 value of 1.5 nM. It is a tricyclic antidepressant used for the research of depression and obsessive-compulsive disorder (OCD).
  • Potent 5-HT reuptake blocker
  • Tricyclic antidepressant
  • Inhibits reuptake of both noradrenaline and 5-HT, with a stronger inhibition of 5-HT reuptake
  • Reduces immobility in the forced swimming test
  • Inhibits marble burying behavior
  • Interferes with autophagic flux and compromises viability of tumorigenic cells
Catalog No. 50-004-08419 Supplier Medchemexpress LLC Supplier No. HYB0457500MG
Clomipramine hydrochloride is a potent 5-HT reuptake blocker with an IC50 value of 1.5 nM. It is a tricyclic antidepressant used for the research of depression and obsessive-compulsive disorder (OCD).
  • Potent 5-HT reuptake blocker
  • Tricyclic antidepressant
  • Inhibits reuptake of both noradrenaline and 5-HT, with a stronger inhibition of 5-HT reuptake
  • Reduces immobility in the forced swimming test
  • Inhibits marble burying behavior
  • Interferes with autophagic flux and compromises viability of tumorigenic cells