Learn More
STEMCELL Technologies ClonaCell™-TCS Medium, Size: 80 mL
SDP
Catalog No. NC1004278
Encompass
Semi-solid methylcellulose-based medium for selection and cloning of suspension-adapted and adherent cells (serum-containing)
Catalog No. NC1004278 Supplier STEMCELL Technologies Supplier No. 03814
Add to Cart
Add to Cart
Semi-solid methylcellulose-based medium for selection and cloning of suspension-adapted and adherent cells (serum-containing)