Learn More
STEMCELL Technologies ClonaCell™-CHO AOF Supplement, Size: 2.5 mL
SDP
Catalog No. NC2046924
Encompass
Animal origin-free, serum-free, L-glutamine-free, and phenol red-free 40X culture supplement for cloning during CHO cell line development
Catalog No. NC2046924 Supplier STEMCELL Technologies Supplier No. 1000382
Animal origin-free, serum-free, L-glutamine-free, and phenol red-free 40X culture supplement for cloning during CHO cell line development