Learn More
STEMCELL Technologies ClonaCell™-HY Medium C, Size: 100 mL
SDP
Catalog No. NC1048700
Encompass
ClonaCell™-HY Medium C is a serum-containing liquid medium used to promote hybridoma viability and recovery after the fusion process (PEG-based or electrofusion) and prior to hypoxanthine, aminopterin, thymidine (HAT) selection. This medium has been verified for use in mouse and rat hybridoma development and reportedly is compatible for production of hybridomas using lymphocytes from a variety of host animals including human, mouse, rat, and hamster.
Catalog No. NC1048700 Supplier STEMCELL Technologies Supplier No. 03803
ClonaCell™-HY Medium C is a serum-containing liquid medium used to promote hybridoma viability and recovery after the fusion process (PEG-based or electrofusion) and prior to hypoxanthine, aminopterin, thymidine (HAT) selection. This medium has been verified for use in mouse and rat hybridoma development and reportedly is compatible for production of hybridomas using lymphocytes from a variety of host animals including human, mouse, rat, and hamster.