Learn More
STEMCELL Technologies ClonaCell™-HY Medium D without HAT, Size: 90 mL
SDP
Catalog No. NC0197802
Encompass
Semi-solid methylcellulose-based hybridoma selection and cloning medium without HAT (serum-containing)
Catalog No. NC0197802 Supplier STEMCELL Technologies Supplier No. 03810
Semi-solid methylcellulose-based hybridoma selection and cloning medium without HAT (serum-containing)