Learn More
STEMCELL Technologies ClonaCell™-HY PEG, Size: 1.5 mL
SDP
Catalog No. NC9069487
Encompass
ClonaCell™-HY PEG is suitable for inducing cellular fusion of myeloma cells with lymphocytes to produce hybridomas. This reagent has been verified for use during mouse and rat hybridoma development and reportedly is compatible for production, cloning, and expansion of hybridomas using lymphocytes from a variety of host animals including human, mouse, rat, and hamster. ClonaCell™-HY PEG is supplied as a 50% (w/v) solution of PEG 3350 in Dulbecco's Modified Eagle's Medium (DMEM) with gentamicin and phenol red. The solution has been pre-tested and qualified for cell fusion.
Catalog No. NC9069487 Supplier STEMCELL Technologies Supplier No. 03806
ClonaCell™-HY PEG is suitable for inducing cellular fusion of myeloma cells with lymphocytes to produce hybridomas. This reagent has been verified for use during mouse and rat hybridoma development and reportedly is compatible for production, cloning, and expansion of hybridomas using lymphocytes from a variety of host animals including human, mouse, rat, and hamster. ClonaCell™-HY PEG is supplied as a 50% (w/v) solution of PEG 3350 in Dulbecco's Modified Eagle's Medium (DMEM) with gentamicin and phenol red. The solution has been pre-tested and qualified for cell fusion.