Learn More
STEMCELL Technologies ClonaCell™-TCS/G418 Kit, Size: 1 Kit
SDP
Catalog No. 501974308
Encompass
This kit is used for selection and cloning applications where the G418 antibiotic is used as the selective agent. ClonaCell™-TCS Medium is a serum-containing, methylcellulose-based semi-solid medium that can be used for selection and cloning of a variety of suspension-adapted and adherent cell lines, such as BHK-1, CHO-K1, CHO-S, CHO-DG44, and HEK-293. This medium contains pre-selected fetal bovine serum (FBS) and bovine serum albumin (BSA). The G418 antibiotic is suitable for use in cell culture for the selection and maintenance of G418-resistant clones when using ClonaCell™-TCS Medium.
Catalog No. 50-197-4308 Supplier STEMCELL Technologies Supplier No. 03822
Add to Cart
Add to Cart
This kit is used for selection and cloning applications where the G418 antibiotic is used as the selective agent. ClonaCell™-TCS Medium is a serum-containing, methylcellulose-based semi-solid medium that can be used for selection and cloning of a variety of suspension-adapted and adherent cell lines, such as BHK-1, CHO-K1, CHO-S, CHO-DG44, and HEK-293. This medium contains pre-selected fetal bovine serum (FBS) and bovine serum albumin (BSA). The G418 antibiotic is suitable for use in cell culture for the selection and maintenance of G418-resistant clones when using ClonaCell™-TCS Medium.