Learn More
STEMCELL Technologies ClonaCell™-TCS/Hygromycin Kit, Size: 1 Kit
SDP
Catalog No. 501974309
Encompass
This kit is meant for selection and cloning applications where the hygromycin B antibiotic is used as the selective agent. ClonaCell™-TCS Medium is a serum-containing, methylcellulose-based semi-solid medium that can be used for selection and cloning of a variety of suspension-adapted and adherent cell lines, such as BHK-1, CHO-K1, CHO-S, CHO-DG44, and HEK-293. This medium contains pre-selected fetal bovine serum (FBS) and bovine serum albumin (BSA). Hygromycin B is suitable for use in cell culture for the selection and maintenance of hygromycin-resistant clones when using ClonaCell™-TCS Medium.
Catalog No. 50-197-4309 Supplier STEMCELL Technologies Supplier No. 03823
Add to Cart
Add to Cart
This kit is meant for selection and cloning applications where the hygromycin B antibiotic is used as the selective agent. ClonaCell™-TCS Medium is a serum-containing, methylcellulose-based semi-solid medium that can be used for selection and cloning of a variety of suspension-adapted and adherent cell lines, such as BHK-1, CHO-K1, CHO-S, CHO-DG44, and HEK-293. This medium contains pre-selected fetal bovine serum (FBS) and bovine serum albumin (BSA). Hygromycin B is suitable for use in cell culture for the selection and maintenance of hygromycin-resistant clones when using ClonaCell™-TCS Medium.