Learn More
ATCC Clone 15 HL-60; Promyelocytic Leukemia; Human (Homo sapiens)
Catalog No. 502382191
Encompass
Clone 15 HL-60 is a cell line exhibiting lymphoblast-like morphology that was isolated from the peripheral blood of a White, 36-year-old, female patient with acute promyelocytic leukemia. This cell line was deposited by S Fischkoff. It can be used in immunology and immune system disorder research.
Catalog No. 50-238-2191 Supplier ATCC Supplier No. CRL1964
Clone 15 HL-60 is a cell line exhibiting lymphoblast-like morphology that was isolated from the peripheral blood of a White, 36-year-old, female patient with acute promyelocytic leukemia. This cell line was deposited by S Fischkoff. It can be used in immunology and immune system disorder research.