Learn More
ATCC clone 1-5c-4; HeLa Contaminant; Human (Homo sapiens)
Catalog No. 502382097
Encompass
The clone 1-5c-4 [Wong-Kilbourne derivative (D) of Chang conjunctiva] cell line was originally thought to be derived from normal conjunctiva, but was subsequently found, based on isoenzyme analysis, HeLa marker chromosomes, and DNA fingerprinting, to have been established via HeLa cell contamination.
Catalog No. 50-238-2097 Supplier ATCC Supplier No. CCL20.2
The clone 1-5c-4 [Wong-Kilbourne derivative (D) of Chang conjunctiva] cell line was originally thought to be derived from normal conjunctiva, but was subsequently found, based on isoenzyme analysis, HeLa marker chromosomes, and DNA fingerprinting, to have been established via HeLa cell contamination.