Learn More
Bio X Cell Monoclonal Antibody, XMG1.2, IFN gamma, 50 MG
Supplier: Bio X Cell BE0055R050MG
The XMG1.2 monoclonal antibody reacts with mouse IFNγ (interferon gamma), a 20 kDa soluble pleiotropic cytokine primarily produced by activated lymphocytes. It exerts immunoregulatory, anti-proliferative, anti-viral, and proinflammatory activities, playing an important role in the activation, growth, and differentiation of various immune cells. The XMG1.2 antibody is a neutralizing antibody.
- Isotype: Rat IgG1, κ
- Formulation: PBS + 0.01% Tween, pH 8.0
- Purity: ≥95% (determined by SDS-PAGE)
- Sterility: 0.2 µm filtration
- Molecular weight: 150 kDa
- Storage: Store at 4°C, do not freeze
- Applications: In vivo and in vitro IFNγ neutralization, ELISPOT, flow cytometry, western blot, in vitro organoids/organ-on-chip
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.