Learn More
Takara Bio CloneAmp™ HiFi PCR Premix, 200 Rxns
SDP
Catalog No. NC0384799
Encompass
A convenient 2X PCR master mix that is optimized for high-fidelity and fast PCR. It is designed for use with Takara's In-Fusion Cloning. CloneAmp HiFi PCR Premix is suitable for high-throughput cloning applications. 200 Rxns
Catalog No. NC0384799 Supplier Takara Bio Supplier No. 639298
A convenient 2X PCR master mix that is optimized for high-fidelity and fast PCR. It is designed for use with Takara's In-Fusion Cloning. CloneAmp HiFi PCR Premix is suitable for high-throughput cloning applications. 200 Rxns