Learn More
STEMCELL Technologies ClonaCell™-HY Medium A, Size: 500 mL
SDP
Catalog No. NC0387703
Encompass
ClonaCell™-HY Medium A is a serum-containing liquid medium formulated for the growth and expansion of parental myeloma cells prior to fusion and for the growth of stable hybridoma clones post-selection. This medium has been verified for use during mouse and rat hybridoma development and reportedly is compatible for production of hybridomas using lymphocytes from a variety of host animals including human, mouse, rat, and hamster.
Catalog No. NC0387703 Supplier STEMCELL Technologies Supplier No. 03801
ClonaCell™-HY Medium A is a serum-containing liquid medium formulated for the growth and expansion of parental myeloma cells prior to fusion and for the growth of stable hybridoma clones post-selection. This medium has been verified for use during mouse and rat hybridoma development and reportedly is compatible for production of hybridomas using lymphocytes from a variety of host animals including human, mouse, rat, and hamster.