Learn More
Agilent Technologies CLONED PFU DNA POLYM
Supplier:  Agilent Technologies 600154
Encompass
Cloned Pfu DNA Polymerase is a recombinant version for high-fidelity PCR and primer-extension reactions. Utilized for PCR cloning and generating blunt-end amplification product. Cloned Pfu DNA polymerase, 10X Cloned Pfu buffer.
Catalog No. 50-125-926