Learn More
Agilent Technologies CLONED PFU DNA POLYM
Catalog No. 50125927
Encompass
Cloned Pfu DNA Polymerase is a recombinant version for high-fidelity PCR and primer-extension reactions. Utilized for PCR cloning and generating blunt-end amplification product. Cloned Pfu DNA polymerase, 10X Cloned Pfu buffer.
Catalog No. 50-125-927 Supplier Agilent Technologies Supplier No. 600159
Cloned Pfu DNA Polymerase is a recombinant version for high-fidelity PCR and primer-extension reactions. Utilized for PCR cloning and generating blunt-end amplification product. Cloned Pfu DNA polymerase, 10X Cloned Pfu buffer.