Learn More
Molecular Devices Molecular Devices CloneMatrix, Semi-Solid Matrix, F/CHO, 40 mL
Catalog No. 5000014482
Encompass_Preferred
CloneMatrix optimized for F/CHO cells is a semi-solid cloning matrix supplied as a 1 x 40 mL preparation that is used to create semi-solid plates for colony formation and single-cell cloning workflows. It is suitable for automated colony picking and manual cloning workflows.
  • Semi-solid matrix for colony formation and cloning.
  • Optimized formulation for F/CHO cells.
  • Supplied as 1 x 40 mL concentrate.
  • Prepares 100 mL final media after preparation.
  • Compatible with automated colony picking systems.
  • Intended for single-cell cloning workflows.
Catalog No. 50-000-14482 Supplier Molecular Devices Supplier No. K8530
CloneMatrix optimized for F/CHO cells is a semi-solid cloning matrix supplied as a 1 x 40 mL preparation that is used to create semi-solid plates for colony formation and single-cell cloning workflows. It is suitable for automated colony picking and manual cloning workflows.
  • Semi-solid matrix for colony formation and cloning.
  • Optimized formulation for F/CHO cells.
  • Supplied as 1 x 40 mL concentrate.
  • Prepares 100 mL final media after preparation.
  • Compatible with automated colony picking systems.
  • Intended for single-cell cloning workflows.