Learn More
Molecular Devices Molecular Devices CloneMatrix Methylcellulose Concentrate, 2.5X, 40 mL
Catalog No. 5000014406
Encompass_Preferred
CloneMatrix 2.5X is a methylcellulose concentrate formulated to convert standard mammalian cell culture media into a semi-solid matrix that supports discrete clonal colony formation and enables automated colony detection and picking.
  • Converts liquid culture media to semisolid media for clonal growth.
  • Supplied as a 2.5x concentrate for dilution to working strength.
  • Packaged as 1 x 40 mL in a 100 mL bottle to minimize aliquoting.
  • Suitable for hybridoma, myeloma, CHO, HEK, and mESC workflows.
  • Preserves positional integrity of colonies for automated screening and picking.
  • Sterile formulation compatible with serum-free media; store refrigerated as specified.
Catalog No. 50-000-14406 Supplier Molecular Devices Supplier No. K8510
CloneMatrix 2.5X is a methylcellulose concentrate formulated to convert standard mammalian cell culture media into a semi-solid matrix that supports discrete clonal colony formation and enables automated colony detection and picking.
  • Converts liquid culture media to semisolid media for clonal growth.
  • Supplied as a 2.5x concentrate for dilution to working strength.
  • Packaged as 1 x 40 mL in a 100 mL bottle to minimize aliquoting.
  • Suitable for hybridoma, myeloma, CHO, HEK, and mESC workflows.
  • Preserves positional integrity of colonies for automated screening and picking.
  • Sterile formulation compatible with serum-free media; store refrigerated as specified.