Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Devices Molecular Devices CloneMedia CHO Growth A Medium, Semi-solid, 90 mL

Catalog No. 5000014438
Encompass_Preferred

CloneMedia CHO Growth A with L-Gln is a semi-solid cell culture medium formulated to support growth and colony formation of suspension-adapted Chinese hamster ovary (CHO) cell lines used in cloning and selection workflows. The product is supplied frozen and is intended for research use only.

  • Semi-solid formulation supplied frozen for stability
  • Formulated for suspension-adapted CHO cells, including DG44 and DUXB11
  • Contains methylcellulose and L-glutamine with recombinant supplements
  • Animal origin free formulation suitable for biopharmaceutical workflows
  • Store at -20°C (up to 6 months); after defrost store at 2-8°C and use within 1 week
  • Manufactured under ISO 9001:2015 quality system, for research use only

Catalog No. 50-000-14438 Supplier Molecular Devices Supplier No. K8800
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CloneMedia CHO Growth A with L-Gln is a semi-solid cell culture medium formulated to support growth and colony formation of suspension-adapted Chinese hamster ovary (CHO) cell lines used in cloning and selection workflows. The product is supplied frozen and is intended for research use only.

  • Semi-solid formulation supplied frozen for stability
  • Formulated for suspension-adapted CHO cells, including DG44 and DUXB11
  • Contains methylcellulose and L-glutamine with recombinant supplements
  • Animal origin free formulation suitable for biopharmaceutical workflows
  • Store at -20°C (up to 6 months); after defrost store at 2-8°C and use within 1 week
  • Manufactured under ISO 9001:2015 quality system, for research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.