Learn More
STEMCELL Technologies CloneR™2, Size: 25 mL
SDP
Catalog No. NC2093379
Encompass
Defined supplement for improving survival of human ES and iPS cells in single-cell workflows
This item is not returnable. View return policy
Catalog No. NC2093379 Supplier STEMCELL Technologies Supplier No. 1000691
Defined supplement for improving survival of human ES and iPS cells in single-cell workflows