Learn More
Cellagen Technology ClonExpress II One Step Cloning Kit
Catalog No. NC1778243
Encompass
A simple, fast, and high efficient cloning technology which is based on a homologous recombination technology. It enables directional insertion of any amplified DNA product into any linearized vector at any site.
Catalog No. NC1778243 Supplier Cellagen Technology Supplier No. C11201
Add to Cart
Add to Cart
A simple, fast, and high efficient cloning technology which is based on a homologous recombination technology. It enables directional insertion of any amplified DNA product into any linearized vector at any site.