Learn More
Cerilliant Corporation Clonidine, 4205-90-7, MFCD00055059, 1mg/mL
Catalog No. NC1246484
Encompass
Empirical Formula (Hill Notation): C9H9Cl2N3, Molecular Weight: 230.09, 1.0 mg/mL in Methanol. Clonidine is a pharmaceutical drug used to treat a wide variety of conditions including hypertension, ADHD, insomnia, chemical withdrawal, and anxiety. Clonidine is often prescribed in combination with other drugs, depending on the clinical condition. This analytical reference standard is suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis and clinical toxicology to pharmaceutical research.
Catalog No. NC1246484 Supplier Cerilliant Corporation Supplier No. C033
Empirical Formula (Hill Notation): C9H9Cl2N3, Molecular Weight: 230.09, 1.0 mg/mL in Methanol. Clonidine is a pharmaceutical drug used to treat a wide variety of conditions including hypertension, ADHD, insomnia, chemical withdrawal, and anxiety. Clonidine is often prescribed in combination with other drugs, depending on the clinical condition. This analytical reference standard is suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis and clinical toxicology to pharmaceutical research.