Learn More
Selleck Chemical LLC Clonidine HCl S2458-1g
SDP
Catalog No. 503112389
Encompass_Preferred
Clonidine HCl(Kapvay) is a direct-acting 2 adrenergic agonist with an ED50 of 0 02 0 01 mg/kg
Catalog No. 50-311-2389 Supplier Selleck Chemical LLC Supplier No. S24581G
Clonidine HCl(Kapvay) is a direct-acting 2 adrenergic agonist with an ED50 of 0 02 0 01 mg/kg