Learn More
Apexbio Technology LLC Clonidine HCl 4205-91-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000088707
Encompass_Preferred
Clonidine Hydrochloride (CAS 4205-91-8) is a synthetic imidazoline derivative that functions as a centrally acting alpha-2 adrenergic receptor agonist By stimulating these receptors in the brainstem clonidine inhibits sympathetic outflow resulting in decreased peripheral vascular resistance and lowered blood pressure In a research context clonidine is frequently used to investigate mechanisms underlying central adrenergic regulation cardiovascular physiology and autonomic nervous system function Its pharmacological profile makes it valuable for studies exploring hypertension neuropharmacology and related therapeutic strategies
Catalog No. 50-000-88707 Supplier Apexbio Technology LLC Supplier No. B13335.1
Clonidine Hydrochloride (CAS 4205-91-8) is a synthetic imidazoline derivative that functions as a centrally acting alpha-2 adrenergic receptor agonist By stimulating these receptors in the brainstem clonidine inhibits sympathetic outflow resulting in decreased peripheral vascular resistance and lowered blood pressure In a research context clonidine is frequently used to investigate mechanisms underlying central adrenergic regulation cardiovascular physiology and autonomic nervous system function Its pharmacological profile makes it valuable for studies exploring hypertension neuropharmacology and related therapeutic strategies