Learn More
Medchemexpress LLC Clonidine (hydrochloride) | 4205-91-8 | 99.9% | 1 G
SDP
Catalog No. 5000408081
Encompass_Preferred
Clonidine hydrochloride is an agonist of α2-adrenoceptor and a potent antihypertensive agent. It has been studied for its various biological activities.
  • Induces CGRP (α and β) mRNA expression in endothelial cells
  • Increases NO levels in endothelial cells
  • Decreases rat body temperature
  • Suppresses dopamine efflux in the prefrontal cortex
  • Reduces blood pressure
  • Enhances RVLM pERK1/2
Catalog No. 50-004-08081 Supplier Medchemexpress LLC Supplier No. HYB0409A1G
Clonidine hydrochloride is an agonist of α2-adrenoceptor and a potent antihypertensive agent. It has been studied for its various biological activities.
  • Induces CGRP (α and β) mRNA expression in endothelial cells
  • Increases NO levels in endothelial cells
  • Decreases rat body temperature
  • Suppresses dopamine efflux in the prefrontal cortex
  • Reduces blood pressure
  • Enhances RVLM pERK1/2