Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Clonidine hydrochloride | 4205-91-8 | 99.9% | 1 ML
SDP

Catalog No. 5000409238
Encompass_Preferred

Clonidine hydrochloride is an agonist of α2-adrenoceptor and a potent antihypertensive agent, primarily intended for research use. This specific product is a 10 mM solution in DMSO.

  • Agonist of α2-adrenoceptor
  • Potent antihypertensive agent
  • For research use only
  • High purity: 99.9%
  • Induces CGRP mRNA expression in endothelial cells
  • Increases NO level in endothelial cells
  • Suppresses dopamine efflux in the prefrontal cortex
  • Solution stable at -20°C for 1 month or -80°C for 6 months

Catalog No. 50-004-09238 Supplier Medchemexpress LLC Supplier No. HYB0409A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Clonidine hydrochloride is an agonist of α2-adrenoceptor and a potent antihypertensive agent, primarily intended for research use. This specific product is a 10 mM solution in DMSO.

  • Agonist of α2-adrenoceptor
  • Potent antihypertensive agent
  • For research use only
  • High purity: 99.9%
  • Induces CGRP mRNA expression in endothelial cells
  • Increases NO level in endothelial cells
  • Suppresses dopamine efflux in the prefrontal cortex
  • Solution stable at -20°C for 1 month or -80°C for 6 months

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.