Learn More
Medchemexpress LLC Clonidine hydrochloride | 4205-91-8 | 99.9% | 5 G
SDP
Catalog No. 5000405299
Encompass_Preferred
Clonidine hydrochloride is an agonist of α2-adrenoceptor and a potent antihypertensive agent. It is for research use only.
  • Agonist of α2-adrenoceptor
  • Potent antihypertensive agent
  • Significantly induces CGRP (α and β) mRNA expression in endothelial cells
  • Significantly increases NO level in endothelial cells
  • Causes a significant decrease in rat body temperature
  • Potently suppresses dopamine efflux in the prefrontal cortex
Catalog No. 50-004-05299 Supplier Medchemexpress LLC Supplier No. HYB0409A5G
Add to Cart
Add to Cart
Clonidine hydrochloride is an agonist of α2-adrenoceptor and a potent antihypertensive agent. It is for research use only.
  • Agonist of α2-adrenoceptor
  • Potent antihypertensive agent
  • Significantly induces CGRP (α and β) mRNA expression in endothelial cells
  • Significantly increases NO level in endothelial cells
  • Causes a significant decrease in rat body temperature
  • Potently suppresses dopamine efflux in the prefrontal cortex